siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 6 | 3065 | |||
| NR_047521.1 | 5 | 2912 | ||||
| NR_047523.1 | 4 | 2816 | ||||
| NR_047524.1 | 4 | 2728 | ||||
| NR_047525.1 | 5 | 2779 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01128| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 6 | 3065 | |||
| NR_047521.1 | 5 | 2912 | ||||
| NR_047523.1 | 4 | 2816 | ||||
| NR_047524.1 | 4 | 2728 | ||||
| NR_047525.1 | 5 | 2779 | ||||