siRNA Details
| Gene Symbol: RNF219-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047001.1 | 6 | 1202 | |||
| GenBank mRNA | AK095779.1 | - | 6 | 1203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF219-AS1| Gene Symbol: RNF219-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047001.1 | 6 | 1202 | |||
| GenBank mRNA | AK095779.1 | - | 6 | 1203 | ||