siRNA Details
| Gene Symbol: LOC101927588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125420.1 | 3 | 636 | |||
| GenBank mRNA | BC015129.1 | - | 3 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927588| Gene Symbol: LOC101927588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125420.1 | 3 | 636 | |||
| GenBank mRNA | BC015129.1 | - | 3 | 640 | ||