siRNA Details
| Gene Symbol: LOC200772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033841.1 | 9 | 1437 | |||
| GenBank mRNA | BC040629.1 | - | 9 | 1437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC200772| Gene Symbol: LOC200772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033841.1 | 9 | 1437 | |||
| GenBank mRNA | BC040629.1 | - | 9 | 1437 | ||