siRNA Details
| Gene Symbol: UCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015379.3 | 3 | 1450 | |||
| GenBank mRNA | AK092401.1 | - | 3 | 1436 | ||
| GU357550.1 | - | 3 | 1302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCA1| Gene Symbol: UCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015379.3 | 3 | 1450 | |||
| GenBank mRNA | AK092401.1 | - | 3 | 1436 | ||
| GU357550.1 | - | 3 | 1302 | |||