siRNA Details
| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024470.1 | 1 | 182 | |||
| GenBank mRNA | BC025345.1 | - | 1 | 182 | 4862156 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO4A1-AS1| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024470.1 | 1 | 182 | |||
| GenBank mRNA | BC025345.1 | - | 1 | 182 | 4862156 | |