siRNA Details
Gene Symbol: LOC101928509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125896.1 | 3 | 310 | |||
GenBank mRNA | AK125257.1 | - | 3 | 310 | ||
DA617431.1 | - | 4 | 556 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA609976.1 | - | 3 | 310 | Chr. 4 - 174097641 - 174220398 [-] on Build GRCh38 |
