siRNA Details
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513404.2 | XP_011511706.1 | 5 | 2237 | ||
GenBank mRNA | AK093972.1 | - | 1 | 2155 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513404.2 | XP_011511706.1 | 5 | 2237 | ||
GenBank mRNA | AK093972.1 | - | 1 | 2155 |