siRNA Details
| Gene Symbol: LINC01587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126517.1 | 3 | 846 | |||
| NR_126518.1 | 3 | 825 | ||||
| NR_126519.1 | 4 | 734 | ||||
| GenBank mRNA | D82070.1 | - | 3 | 823 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01587| Gene Symbol: LINC01587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126517.1 | 3 | 846 | |||
| NR_126518.1 | 3 | 825 | ||||
| NR_126519.1 | 4 | 734 | ||||
| GenBank mRNA | D82070.1 | - | 3 | 823 | ||