siRNA Details
| Gene Symbol: LOC102724532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135480.1 | 1 | 26 | |||
| GenBank mRNA | HY017353.1 | - | 1 | 27 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK004660.1 | - | 1 | 25 | Chr. 17 - 47891233 - 47895812 [-] on Build GRCh38 | |

LOC102724532
Genomic Map