siRNA Details
| Gene Symbol: LOC102724532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135480.1 | 2, 3 | 167 | |||
| GenBank mRNA | HY017353.1 | - | 2, 3 | 168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724532| Gene Symbol: LOC102724532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135480.1 | 2, 3 | 167 | |||
| GenBank mRNA | HY017353.1 | - | 2, 3 | 168 | ||