siRNA Details
| Gene Symbol: LOC100996842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136401.1 | 1 | 129 | |||
| NR_136402.1 | 1 | 129 | ||||
| GenBank mRNA | BM888151.1 | - | 1 | 118 | ||
| HY076064.1 | - | 1 | 130 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996842| Gene Symbol: LOC100996842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136401.1 | 1 | 129 | |||
| NR_136402.1 | 1 | 129 | ||||
| GenBank mRNA | BM888151.1 | - | 1 | 118 | ||
| HY076064.1 | - | 1 | 130 | |||