siRNA Details
| Gene Symbol: LOC101927040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110954.1 | 6 | 885 | |||
| GenBank mRNA | AK055332.1 | - | 6 | 887 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927040| Gene Symbol: LOC101927040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110954.1 | 6 | 885 | |||
| GenBank mRNA | AK055332.1 | - | 6 | 887 | ||