siRNA Details
| Gene Symbol: LOC102723526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134271.1 | 2 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723526| Gene Symbol: LOC102723526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134271.1 | 2 | 456 | |||