siRNA Details
Gene Symbol: LOC102723526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134270.1 | 1 | 15 | |||
NR_134271.1 | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA675095.1 | - | 1 | 16 | Chr. 5 - 17444010 - 17483946 [+] on Build GRCh38 |
