siRNA Details
| Gene Symbol: LOC101926964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110628.1 | 6 | 520 | |||
| GenBank mRNA | AK097193.1 | - | 6 | 523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926964| Gene Symbol: LOC101926964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110628.1 | 6 | 520 | |||
| GenBank mRNA | AK097193.1 | - | 6 | 523 | ||