siRNA Details
| Gene Symbol: EFCAB6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046563.1 | 3 | 1555 | |||
| GenBank mRNA | AK090843.1 | - | 3 | 1555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFCAB6-AS1| Gene Symbol: EFCAB6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046563.1 | 3 | 1555 | |||
| GenBank mRNA | AK090843.1 | - | 3 | 1555 | ||