siRNA Details
| Gene Symbol: LOC100507477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121622.1 | 4 | 1015 | |||
| NR_121623.1 | 2 | 496 | ||||
| GenBank mRNA | BC038188.1 | - | 4 | 1004 | ||
| BU160297.1 | - | 2 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507477| Gene Symbol: LOC100507477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121622.1 | 4 | 1015 | |||
| NR_121623.1 | 2 | 496 | ||||
| GenBank mRNA | BC038188.1 | - | 4 | 1004 | ||
| BU160297.1 | - | 2 | 496 | |||