siRNA Details
| Gene Symbol: LOC105369920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135016.1 | 1 | 46 | |||
| GenBank mRNA | AI792597.1 | - | 1 | 46 | ||
| BX108644.1 | - | 1 | 46 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369920| Gene Symbol: LOC105369920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135016.1 | 1 | 46 | |||
| GenBank mRNA | AI792597.1 | - | 1 | 46 | ||
| BX108644.1 | - | 1 | 46 | |||