siRNA Details
| Gene Symbol: A1BG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015380.2 | 3 | 1304 | |||
| GenBank mRNA | AK027222.1 | - | 3 | 1304 | ||
| BC040926.1 | - | 3 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A1BG-AS1| Gene Symbol: A1BG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015380.2 | 3 | 1304 | |||
| GenBank mRNA | AK027222.1 | - | 3 | 1304 | ||
| BC040926.1 | - | 3 | 381 | |||