siRNA Details
| Gene Symbol: LOC100506271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110515.1 | 1 | 1539 | |||
| GenBank mRNA | AK097787.1 | - | 1 | 1341 | ||
| BC043218.2 | - | 1 | 1539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506271| Gene Symbol: LOC100506271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110515.1 | 1 | 1539 | |||
| GenBank mRNA | AK097787.1 | - | 1 | 1341 | ||
| BC043218.2 | - | 1 | 1539 | |||