siRNA Details
| Gene Symbol: LINC01447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108090.1 | 2 | 760 | |||
| GenBank mRNA | AK056440.1 | - | 2 | 760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01447| Gene Symbol: LINC01447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108090.1 | 2 | 760 | |||
| GenBank mRNA | AK056440.1 | - | 2 | 760 | ||