siRNA Details
| Gene Symbol: LOC100507661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109989.1 | 5 | 577 | |||
| GenBank mRNA | BX118401.1 | - | 5 | 577 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK824549.1 | - | 4 | 505 | Chr. 3 - 168903404 - 168921842 [+] on Build GRCh38 | |

LOC100507661
Genomic Map