siRNA Details
| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015359.1 | 4 | 714 | |||
| GenBank mRNA | AY250185.1 | - | 7 | 1237 | ||
| BC019264.1 | - | 7 | 790 | |||
| BC033127.1 | - | 4 | 695 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL523811.3 | - | 4 | 714 | Chr. 4 - 84966818 - 85006497 [+] on Build GRCh38 | |

WDFY3-AS2
Genomic Map