siRNA Details
| Gene Symbol: C17orf102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207454.2 | NP_997337.2 | 2 | 3430 | ||
| GenBank mRNA | AK126768.1 | - | 2 | 3430 | ||
| BC142651.1 | - | 2 | 3414 | 40148022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17orf102| Gene Symbol: C17orf102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207454.2 | NP_997337.2 | 2 | 3430 | ||
| GenBank mRNA | AK126768.1 | - | 2 | 3430 | ||
| BC142651.1 | - | 2 | 3414 | 40148022 | ||