siRNA Details
| Gene Symbol: LINC00922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027755.1 | 9 | 872 | |||
| GenBank mRNA | BC037902.1 | - | 9 | 878 | ||
| BC104446.1 | - | 10 | 880 | |||
| BC104447.1 | - | 6 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00922| Gene Symbol: LINC00922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027755.1 | 9 | 872 | |||
| GenBank mRNA | BC037902.1 | - | 9 | 878 | ||
| BC104446.1 | - | 10 | 880 | |||
| BC104447.1 | - | 6 | 481 | |||