siRNA Details
| Gene Symbol: LINC00923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025311.1 | - | 3 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00923| Gene Symbol: LINC00923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025311.1 | - | 3 | 1678 | ||