siRNA Details
| Gene Symbol: FLJ37505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033987.1 | 3 | 2000 | |||
| GenBank mRNA | AK094824.1 | - | 3 | 1969 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ37505| Gene Symbol: FLJ37505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033987.1 | 3 | 2000 | |||
| GenBank mRNA | AK094824.1 | - | 3 | 1969 | ||