siRNA Details
| Gene Symbol: LOC101929607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110039.1 | 3 | 1237 | |||
| GenBank mRNA | BC042024.1 | - | 2 | 1236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929607| Gene Symbol: LOC101929607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110039.1 | 3 | 1237 | |||
| GenBank mRNA | BC042024.1 | - | 2 | 1236 | ||