siRNA Details
| Gene Symbol: NRG1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037250.1 | - | 4 | 599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NRG1-IT1| Gene Symbol: NRG1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037250.1 | - | 4 | 599 | ||