siRNA Details
| Gene Symbol: ALMS1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046762.1 | 2 | 474 | |||
| GenBank mRNA | BC041487.1 | - | 2 | 481 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM451564.1 | - | 480 | Chr. - - [-] on Build | ||

ALMS1-IT1
Genomic Map