siRNA Details
Gene Symbol: LOC255654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040002.1 | 3 | 864 | |||
GenBank mRNA | BC039539.1 | - | 3 | 863 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC255654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040002.1 | 3 | 864 | |||
GenBank mRNA | BC039539.1 | - | 3 | 863 |