siRNA Details
| Gene Symbol: UMODL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027243.1 | 2 | 1493 | |||
| GenBank mRNA | AK090790.1 | - | 2 | 1407 | ||
| BC063412.1 | - | 2 | 1410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UMODL1-AS1| Gene Symbol: UMODL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027243.1 | 2 | 1493 | |||
| GenBank mRNA | AK090790.1 | - | 2 | 1407 | ||
| BC063412.1 | - | 2 | 1410 | |||