siRNA Details
Gene Symbol: OTX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029385.1 | 4 | 855 | |||
GenBank mRNA | AB593040.1 | - | 3 | 845 | ||
BC041486.2 | - | 4 | 854 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OTX2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029385.1 | 4 | 855 | |||
GenBank mRNA | AB593040.1 | - | 3 | 845 | ||
BC041486.2 | - | 4 | 854 |