siRNA Details
| Gene Symbol: LINC00893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027455.5 | 10 | 1624 | |||
| GenBank mRNA | AK124040.1 | - | 10 | 1624 | ||
| U66044.1 | - | 4 | 385 | |||
| U66045.1 | - | 4 | 385 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI239869.1 | - | 3 | 271 | Chr. X - 149528168 - 149529680 [-] on Build GRCh38 | |

LINC00893
Genomic Map