siRNA Details
Gene Symbol: LINC01485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108028.1 | 4 | 497 | |||
GenBank mRNA | CB162805.1 | - | 4 | 480 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB163354.1 | - | 4 | 480 | Chr. 5 - 173786947 - 173790925 [-] on Build GRCh38 |
