siRNA Details
| Gene Symbol: LOC100505824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135559.1 | 1 | 250 | |||
| GenBank mRNA | BC033999.2 | - | 1 | 251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505824| Gene Symbol: LOC100505824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135559.1 | 1 | 250 | |||
| GenBank mRNA | BC033999.2 | - | 1 | 251 | ||