siRNA Details
| Gene Symbol: LOC100505942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104656.1 | 1 | 293 | |||
| GenBank mRNA | HY127858.1 | - | 1 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505942| Gene Symbol: LOC100505942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104656.1 | 1 | 293 | |||
| GenBank mRNA | HY127858.1 | - | 1 | 293 | ||