siRNA Details
| Gene Symbol: TTLL10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125828.1 | - | 1 | 2697 | ||
| AK128833.1 | - | 2 | 2073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL10-AS1| Gene Symbol: TTLL10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125828.1 | - | 1 | 2697 | ||
| AK128833.1 | - | 2 | 2073 | |||