siRNA Details
Gene Symbol: LOC101929147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125974.1 | 2 | 417 | |||
NR_125975.1 | 4 | 601 | ||||
NR_125976.1 | 4 | 601 | ||||
NR_125977.1 | 3 | 534 | ||||
GenBank mRNA | AK125759.1 | - | 4 | 287 | ||
BC034623.1 | - | 4 | 267 | |||
BC043601.1 | - | 2 | 388 |
