siRNA Details
| Gene Symbol: LOC100505915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125434.1 | 3 | 817 | |||
| GenBank mRNA | AK125313.1 | - | 3 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505915| Gene Symbol: LOC100505915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125434.1 | 3 | 817 | |||
| GenBank mRNA | AK125313.1 | - | 3 | 817 | ||