siRNA Details
| Gene Symbol: LOC101928053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120540.1 | 3 | 2180 | |||
| GenBank mRNA | AK056207.1 | - | 3 | 2177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928053| Gene Symbol: LOC101928053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120540.1 | 3 | 2180 | |||
| GenBank mRNA | AK056207.1 | - | 3 | 2177 | ||