siRNA Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092080.1 | - | 7 | 828 | ||
| AK092150.1 | - | 4 | 503 | |||
| AK092820.1 | - | 1 | 1218 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA690246.1 | - | 4 | 504 | Chr. 19 - 24033445 - 24065626 [+] on Build GRCh38 | |

ZNF254
Genomic Map