siRNA Details
| Gene Symbol: LOC403323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122076.1 | 2 | 595 | |||
| NR_122077.1 | 2 | 705 | ||||
| GenBank mRNA | AK000451.1 | - | 2 | 705 | ||
| AK309896.1 | - | 2 | 215 | |||
| DB521113.1 | - | 2 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC403323| Gene Symbol: LOC403323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122076.1 | 2 | 595 | |||
| NR_122077.1 | 2 | 705 | ||||
| GenBank mRNA | AK000451.1 | - | 2 | 705 | ||
| AK309896.1 | - | 2 | 215 | |||
| DB521113.1 | - | 2 | 246 | |||