siRNA Details
| Gene Symbol: DLEU7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046551.1 | 5 | 1501 | |||
| GenBank mRNA | BC035769.1 | - | 5 | 1511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLEU7-AS1| Gene Symbol: DLEU7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046551.1 | 5 | 1501 | |||
| GenBank mRNA | BC035769.1 | - | 5 | 1511 | ||