siRNA Details
Gene Symbol: LINC01358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110626.2 | 1, 2 | 150 | |||
GenBank mRNA | HY083857.1 | - | 1, 2 | 150 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110626.2 | 1, 2 | 150 | |||
GenBank mRNA | HY083857.1 | - | 1, 2 | 150 |