siRNA Details
Gene Symbol: LOC100507388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121617.1 | 4 | 1350 | |||
GenBank mRNA | AK027257.1 | - | 4 | 1351 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121617.1 | 4 | 1350 | |||
GenBank mRNA | AK027257.1 | - | 4 | 1351 |