siRNA Details
| Gene Symbol: MIR137HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046105.1 | 5 | 1476 | |||
| GenBank mRNA | AK094607.1 | - | 5 | 1388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR137HG| Gene Symbol: MIR137HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046105.1 | 5 | 1476 | |||
| GenBank mRNA | AK094607.1 | - | 5 | 1388 | ||