siRNA Details
Gene Symbol: LINC00608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015390.1 | 1 | 180 | |||
NR_024385.1 | 1 | 180 | ||||
GenBank mRNA | AK098539.1 | - | 1 | 180 | ||
BC033552.1 | - | 1 | 162 | |||
BC049373.1 | - | 1 | 180 | |||
BC121084.1 | - | 1 | 162 | |||
BC121085.1 | - | 1 | 162 |
