siRNA Details
Gene Symbol: LOC100506551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103809.1 | 1 | 313 | |||
GenBank mRNA | AK127077.1 | - | 1 | 314 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103809.1 | 1 | 313 | |||
GenBank mRNA | AK127077.1 | - | 1 | 314 |